3,5-Di-O-caffeoylquinic acid ≥97% (by HPLC)
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
Synonyms: Isochlorogenic acid, (1R,3R,4S,5R)-3,5-Bis((3-(3,4-dihydroxyphenyl)acryloyl)oxy)-1,4-dihydroxy-3-methylcyclohexane-1-carboxylic acid, 3,5-CQA, Isochlorogenic acid A
CAS Number:2450-53-5
:
Formula:C₂₅H₂₄O₁₂
MDL Number:MFCD00951290
Molecular Weight:516.5 g/mol
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Purity:≥97% by HPLC
- Identity:Determined by ¹H-NMR, ¹³C-NMR and MS
- Source:Isolated from Cynara scolymus
Specification Test Results
Frequently Bought Together