Order Entry
Ireland
ContactUsLinkComponent
Water Reag. Ph. Eur. 1095507 particle-free
Water Reag. Ph. Eur. 1095507 particle-free
  87962.290
 :  
undefined
Water Reag. Ph. Eur. 1095507 particle-free
  87962.290
 :  
  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
 : Hydrogen oxide
 :
 :18.02 g/mol
 :100
 :0
 :1 g/cm³ (4 °C)
 :Ambient
 :231-791-2
 :7732-18-5
 :H₂O
 :MFCD00011332
 :13,10098