Specifications
- Description:WarmMark ascending time-temperature indicator (–18 ℃/0 ℉, 12 hours), 46×19×1,5 mm
- Environmentally Preferable:
- Accuracy:±1 °C/±2 °F
- Pk:1
Specifications
About this item
WarmMark is time-temperature indicator sticker, that provides insights into your cold chain logistics so you will know which packages may have experienced thermal damage and which were handled properly.
Monitor products like vaccines, injectables and any other element transported in dry ice. It is a single-use, ascending time-temperature indicator which alerts users of exposure to unacceptable temperature conditions.
This single-use temperature recorder for shipping turns red when it reaches the temperature threshold you selected. Then, unlike many temperature indicator stickers, WarmMark starts a countdown, with its three individual indicator dots changing color for brief, moderate, and prolonged exposure. Those durations vary by temperature. So, for example, 'brief' exposure means 2 hours at 0 °C/32 °F, but means 30 minutes at 25 °C/77 °F and above. Based on the temperature and exposure, you’ll know whether a product remains safe and effective or whether it should be replaced.
As products become increasingly temperature sensitive, your ability to gain insights into your cold chain management and logistics becomes vital. WarmMark provides those insights, giving you hard data to support your decisions about today’s shipments and about your cold chain logistics going forward.
Store in a dry environment below response temperature for optimal shelf life.