Order Entry
Ireland
ContactUsLinkComponent
Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one 97%, pure
Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one 97%, pure
Certificates
undefined
Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one 97%, pure

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4875 to 1.4895 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together