3-(2-Hydroxyethyl)indole 97%
: 3-Indoleethanol, Indol-3-ethanol, Tryptophol, β-Indol-3-ylethanol
:526-55-6
:
:C₁₀H₁₁NO
:MFCD00005659
:174 °C (2.7mbar)
:161.2 g/mol
:57…60 °C
:255 °C
:12,09930
:208-393-2
- Environmentally Preferable:
- Appearance:Off-white to brown Powder or crystals or flakes or crystalline mass
- Infrared spectrum:Conforms
- Melting point:56 to 61 °C
- GC:≥96.0 %
Frequently Bought Together