Order Entry
Ireland
ContactUsLinkComponent
Cyanine 5 maleimide ≥90% [equivalent to Cy5® maleimide]
Cyanine 5 maleimide ≥90% [equivalent to Cy5® maleimide]
Catalog # AATB152
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cyanine 5 maleimide ≥90% [equivalent to Cy5® maleimide]
Catalog # AATB152
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Cy5 maleimide
 :
 :892.96 g/mol