Test tubes with side tube
DURAN® borosilicate glass, round bottom, beaded rim.
Test tubes with side tube (35×8 mm Ø) positioned 20 mm below rim.
Frequently Bought Together
DURAN® borosilicate glass, round bottom, beaded rim.
Test tubes with side tube (35×8 mm Ø) positioned 20 mm below rim.