Airtight, Water-Resistant Storage Boxes
Supplier: MULTIROIR
Certificates
About this item
These airtight boxes are designed to withstand the harshest conditions, both indoors and outdoors. When tightly closed, the boxes become completely sealed, effectively preventing moisture and dust from reaching the contents inside.
- Perfect waterproofing (IP 64/IP 44 standards)
- Nestable and stackable
- Resistant to temperature variations (−40 to 70 °C)
- Chemical-resistant
- Manufactured from polypropylene
- Suitable for food contact
- Compliant with 600×400 and 400×300 palletisation standards
Frequently Bought Together