Designed to determine the protein content of food.
- Independent heat controller lets you adjust the temperature range of each mantle individually
- Stainless steel construction is rugged and easy to clean
Run several projects simultaneously using these multi-position Kjeldahl extraction heaters. Heaters are available with either two or six individually controlled heating mantles that accommodate a wide range of round-bottom flask options. Back-mounted brackets hold flask support rods up to 12,7 mm in diameter.
Each mantle has an independent heater with control dial, on/off switch, and indicator light, and operates to a maximum element temperature of 800 °C. A quartz heating element is housed in a thermally insulated cartridge for easy replacement, extending the life of the heater. Heat controllers located in the lower part of the unit is safely separated from the upper heating elements by a stainless steel tray and well-ventilated air space. Extra safety features include an earth screen and double fuses on the supply line.
Use fume collection tube with micro-Kjeldahl extraction heaters to safely collect and exhaust fumes.
- Catalog No:
- ELET03075-22
- ELET03075-16
- ELET03075-20
- ELET03075-12
- ELET03075-14
- ELET03075-08
- ELET03075-10
- ELET03075-02
- ELET03075-04
- ELET03074-34
- Electrical:
- 230 VAC, 50/60 Hz
- 230 VAC, 50/60 Hz
- 230 VAC, 50/60 Hz
- 115 VAC, 50/60 Hz
- 230 VAC, 50/60 Hz
- 230 VAC, 50/60 Hz
- 230 VAC, 50/60 Hz
- 230 VAC, 50/60 Hz
- 230 VAC, 50/60 Hz
- 230 VAC, 50/60 Hz
- Max. temperature:
- 800 ℃
- 800 ℃
- 800 ℃
- 800 ℃
- 800 ℃
- 800 ℃
- 800 ℃
- 800 ℃
- 800 ℃
- 800 ℃