Order Entry
Ireland
ContactUsLinkComponent
Anti-GP210 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-GP210 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-13495R-A680
 :  Bioss
Anti-GP210 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-13495R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    GP210
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    1161-1260/1887
  • Form:
    Liquid
  • Gene ID:
    23225
  • Antigen synonyms:
    Nucleoporin 210|Nucleoporin210|Pore membrane protein of 210 kDa|NUP210|FLJ22389|Nucleoporin Nup210|KIAA0906|PO210_HUMAN|Nuclear pore protein gp210|Nucleoporin 210kDa|Nuclear envelope pore membrane protein POM 210|Nuclear pore membrane glycoprotein 210|GP 210|POM210|NUP 210|POM 210
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    203 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human GP210
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

The nuclear pore complex (NPC) mediates bidirectional macromolecular traffic between the nucleus and cytoplasm in eukaryotic cells and is comprised of more than 100 different subunits. Many of the subunits belong to a family called nucleoporins (Nups), which are characterized by the presence of O-linked-N-acetylglucosamine moieties and a distinctive pentapeptide repeat (XFXFG). gp210, also known as Nup210 (nucleoporin 210kDa) or POM210, is a 1,887 amino acid single-pass type I membrane protein that localizes to both the endoplasmic reticulum and to the nucleus, specifically within the NPC. Expressed ubiquitously with highest expression in pancreas, testis, lung, ovary and liver, gp210 functions as a nucleoporin that is capable of dimerization and is essential for the assembly, fusion and structural integrity of the NPC. gp210 exists as multiple alternatively spliced isoforms and is subject to post-translational phosphorylation.

Type: Primary
Antigen: GP210
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1161-1260/1887
Host: Rabbit
Isotype: IgG
Reactivity: