Anti-PAICS Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-6369R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:PAICS
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Environmentally Preferable:
- Epitope:201-300/425
- Form:Liquid
- Gene ID:10606
- Antigen synonyms:PUR6_HUMAN|SAICAR synthetase|MGC5024|AIR carboxylase|ADE2|MGC1343|ADE2H1|PAIS|DKFZp781N1372|Phosphoribosylaminoimidazole carboxylase|Paics|AIRC
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:47 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human PAICS
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
This gene encodes a bifunctional enzyme containing phosphoribosylaminoimidazole carboxylase activity in its N-terminal region and phosphoribosylaminoimidazole succinocarboxamide synthetase in its C-terminal region. It catalyzes steps 6 and 7 of purine biosynthesis. The gene is closely linked and divergently transcribed with a locus that encodes an enzyme in the same pathway, and transcription of the two genes is coordinately regulated. The human genome contains several pseudogenes of this gene. Multiple transcript variants encoding different isoforms have been found for this gene.
Type: Primary
Antigen: PAICS
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/425
Host: Rabbit
Isotype: IgG
Reactivity: