Antistatic Waste Bags
: DESCO
Antistatic waste bags are non-tribocharging and are designed for use in ESD protected areas where electrostatic sensitive devices are present. Even at low humidity they do not become charged with static electricity.
- Made of antistatic polyethylene
- Charges to <100 V, T1000: < 4 sec
- Comparable in strength to conventional refuse sacks
- Printed in black with ESD protective symbol
They are made from high quality polyethylene and are as strong as conventional refuse sacks.
All sizes are printed with the ESD protective symbol per EN 61340-5.
Frequently Bought Together