Manual Tubing Pinch Valves, Delrin®
Supplier: Avantor Fluid Handling
Certificates
About this item
An ideal option to secure tubing.
- Constructed of durable Delrin®
Fluid contacts only the tubing—ideal for use in pharmaceutical and food industries where eliminating contamination is critical. Obtain precise and repeatable flow rates using easy-to-read metered scale on valve body. Valves are made of Delrin®. Tubing is not included—order flexible tubing with durometer (Shore A) of 60 or less from our "Tubing" section.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX98002-20
- MFLX98002-10
- MFLX98002-00
- Pinch valve tube size:
- 7/16" to 1/2" OD
- 5/16" to 3/8" OD
- 5/32" to 1/4" OD
- Warranty:
- 3 Months
- 3 Months
- 3 Months