Order Entry
Ireland
ContactUsLinkComponent
+GF+ Signet Rotor-X Paddle-Wheel Flow Sensors
+GF+ Signet Rotor-X Paddle-Wheel Flow Sensors
Certificates
undefined
+GF+ Signet Rotor-X Paddle-Wheel Flow Sensors
Specifications
  • Max. operating temperature:
  • 100 °C (212 °F)
  • 85 °C (185 °F)
  • 100 °C (212 °F)
  • Power:
  • 5 - 24 VDC
  • 5 - 24 VDC
  • 5 - 24 VDC
  • Repeatability:
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • Wetted materials:
  • PVDF, Hastelloy C
  • Polypropylene, Titanium
  • Polypropylene, Titanium
  • Warranty:
  • 1 year part and 2 years labor
  • 1 year part and 2 years labor
  • 1 year part and 2 years labor