3-(Trifluoromethyl)isonicotinic acid 99%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 3-(Trifluoromethyl)pyridine-4-carboxylic acid
CAS Number:590371-38-3
:
Formula:C₇H₄F₃NO₂
MDL Number:MFCD08234940
Density:0.939 g/cm³ (25 °C)
Molecular Weight:191.11 g/mol
Melting Pt:212…215 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together