5-(Chloromethyl)-2-(trifluoromethyl)pyridine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:386715-33-9
:
Formula:C₇H₅ClF₃N
MDL Number:MFCD01862650
Boiling Pt:92…94 °C/6…8mmHg
Molecular Weight:195.57 g/mol
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together