2,4-Dimethoxy-5-fluoroaniline
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 5-Fluoro-2,4-dimethoxyaniline
CAS Number:195136-65-3
:
Formula:C₈H₁₀FNO₂
MDL Number:MFCD12547861
Molecular Weight:171.17 g/mol
Melting Pt:47…49 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1 III
:GHS06
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together