1-(3-Fluorophenyl)biguanide hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2267-49-4
:
Formula:C₈H₁₁ClFN₅
MDL Number:MFCD00203529
Melting Pt:231…235 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together