Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chloro-3-fluoro-2-methoxypyridine
5-Chloro-3-fluoro-2-methoxypyridine 98%
5-Chloro-3-fluoro-2-methoxypyridine 98%
APOSPC408037-5G
:
Apollo Scientific
:
5-Chloro-3-fluoro-2-methoxypyridine 98%
APOSPC408037-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
Environmentally Preferable:
:
2-Methoxy-5-chloro-3-fluoropyridine
:
:
161.56 g/mol
:
Ambient
:
886374-01-2
:
C₆H₅ClFNO
:
MFCD07375056