5'-Fluoro-2'-hydroxyacetophenone 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 1-(5-Fluoro-2-hydroxyphenyl)ethanone
CAS Number:394-32-1
:
Formula:C₈H₇FO₂
MDL Number:MFCD00011668
Boiling Pt:65…66 °C/8mmHg
Molecular Weight:154.14 g/mol
Melting Pt:56…58 °C
EINECS:206-893-5
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together