5-(Ethylcarbamoyl)-2-fluorophenylboronic acid 97%
Catalog # APOSPC3528-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: (5-(Ethylcarbamoyl)-2-fluorophenyl)boronic acid
:
:211.0000 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:874289-45-9
Formula:C₉H₁₁BFNO₃
MDL Number:MFCD08436027
Melting Pt:141…143 °C