3,5-Bis(trifluoromethyl)benzenesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 3,5-Bis(trifluoromethyl)benzene-1-sulphonyl chloride
CAS Number:39234-86-1
:
Formula:C₈H₃ClF₆O₂S
MDL Number:MFCD00014725
Molecular Weight:312.62 g/mol
Melting Pt:34…38 °C
Flash Pt:108 °C
Storage Temperature:Ambient
UN:3261
ADR:8 II
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together