3-Fluoro-2-methoxybenzonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:77801-22-0
:
Formula:C₈H₆FNO
MDL Number:MFCD06637312
Molecular Weight:151.14 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together