4-Bromo-1-(bromomethyl)-2-chlorobenzene 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:89720-77-4
:
Formula:C₇H₅Br₂Cl
MDL Number:MFCD15526905
Molecular Weight:284.3756 g/mol
UN:3265
ADR:8 III
:GHS05
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together