3-(Methylsulphonyl)-1-propanol 95%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 3-(Methylsulphonyl)propan-1-ol
CAS Number:2058-49-3
:
Formula:C₄H₁₀O₃S
MDL Number:MFCD01696727
Molecular Weight:138.19 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1 III
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together