5-Ethyl-1-methyl-1H-pyrazole-3-carboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:165744-15-0
:
Formula:C₇H₁₀N₂O₂
MDL Number:MFCD04115075
Molecular Weight:154.17 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together