N,N,N',N'-Tetramethyl-2-butene-1,4-diamine 95%
Catalog # APOSOR925517-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
Synonyms: N,N,N',N'-Tetramethyl-1,4-diamino-2-butene
:
:142.25 g/mol
Boiling Pt:171…172 °C
Density:0.808 g/cm³ (25 °C)
Flash Pt:51 °C
Storage Temperature:Ambient
UN:2734
CAS Number:4559-79-9
Formula:C₈H₁₈N₂
MDL Number:MFCD00008336
ADR:8+3 II