Order Entry
Ireland
ContactUsLinkComponent
3-(1H-1,2,4-Triazol-1-yl)benzaldehyde 97%
3-(1H-1,2,4-Triazol-1-yl)benzaldehyde 97%
Catalog # APOSOR9235-250MG
Supplier:  Apollo Scientific
CAS Number:  
3-(1H-1,2,4-Triazol-1-yl)benzaldehyde 97%
Catalog # APOSOR9235-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 3-(1,2,4-Triazol-1-yl)benzaldehyde
 :
 :173.1714 g/mol
UN:1325
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:868755-54-8
Formula:C₉H₇N₃O
MDL Number:MFCD08271896
Melting Pt:114.5…115.5 °C
ADR:4.1 III