5-Chloromethyl-2-(1H-pyrazol-1-yl)pyridine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:748796-39-6
:
Formula:C₉H₈ClN₃
MDL Number:MFCD06658399
Molecular Weight:193.634 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together