2-Amino-5-bromo-3-methylbenzoic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:206548-13-2
:
Formula:C₈H₈BrNO₂
MDL Number:MFCD05664860
Molecular Weight:230.0600 g/mol
Melting Pt:230…240 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together