2,6-Dimethylphenylhydrazine hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: (2,6-Dimethylphenyl)hydrazine hydrochloride
CAS Number:2538-61-6
:
Formula:C₈H₁₃ClN₂
MDL Number:MFCD01861265
Molecular Weight:172.6552 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1 III
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together