2-Chloro-3-cyano-4-methoxypyridine
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2-Chloro-4-methoxynicotinonitrile
CAS Number:98645-43-3
:
Formula:C₇H₅ClN₂O
MDL Number:MFCD00975444
Molecular Weight:168.5804 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1 III
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together