5-(Chloroacetyl)-6-chlorooxindole 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 6-Chloro-5-(chloroacetyl)-1,3-dihydro-2H-indol-2-one
CAS Number:118307-04-3
:
Formula:C₁₀H₇Cl₂NO₂
MDL Number:MFCD04115735
Melting Pt:211…213 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8 III
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together