3,4,5-Trimethylaniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1639-31-2
:
Formula:C₉H₁₃N
MDL Number:MFCD00130022
Molecular Weight:135.21 g/mol
Melting Pt:79…80 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1 III
:GHS06
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together