Order Entry
Ireland
ContactUsLinkComponent
1-Methyl-1H-pyrazol-5-ol 97%
1-Methyl-1H-pyrazol-5-ol 97%
Catalog # APOSOR55337-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Methyl-1H-pyrazol-5-ol 97%
Catalog # APOSOR55337-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 1-Methyl-1H-pyrazol-5-ol, 5-Hydroxy-1-methyl-1H-pyrazole
 :
 :98.1032 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:33641-15-5
Formula:C₄H₆N₂O
MDL Number:MFCD05864418
Melting Pt:110…114 °C