5,5'-Dibromo-2,2'-bithiophene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4805-22-5
:
Formula:C₈H₄Br₂S₂
MDL Number:MFCD00219110
Molecular Weight:324.05 g/mol
Melting Pt:144…146 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together