Bis[2-(diphenylphosphino)ethyl]ammonium chloride
Catalog # APOSOR54422-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: Bis(2-(diphenylphosphino)ethyl)amine hydrochloride
:
:477.953 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:66534-97-2
Formula:C₂₈H₃₀ClNP₂
MDL Number:MFCD00061569
Melting Pt:175…180 °C