1,12-Dibromododecane 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3344-70-5
:
Formula:BrH₂C(CH₂)₁₀CH₂Br
MDL Number:MFCD00000226
Boiling Pt:214…216 °C/15mmHg
Molecular Weight:328.13 g/mol
Melting Pt:37…42 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together