1-Methyl-5-nitro-2,3-dihydro-1H-1,3-benzodiazol-2-one
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 1-Methyl-5-nitro-1,3-dihydro-2H-benzo[d]imidazol-2-one
CAS Number:66108-85-8
:
Formula:C₈H₇N₃O₃
MDL Number:MFCD18157683
Molecular Weight:193.16 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together