Fmoc-Asp-OBzl 95%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: N-Fmoc-L-aspartic acid 1-benzyl ester, Fmoc-Asp-Obzl
CAS Number:86060-83-5
:
Formula:C₂₆H₂₃NO₆
MDL Number:MFCD00198201
Molecular Weight:445.471 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together