4-Methyl-1H-indole-2-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:18474-57-2
:
Formula:C₁₀H₉NO₂
MDL Number:MFCD02664476
Molecular Weight:175.1800 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together