1,2,3,4,5-Pentamethylcyclopentadiene, Technical Grade
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Pentamethylcyclopentadiene, 1,2,3,4,5-Pentamethylcyclopenta-1,3-diene, 1,2,3,4,5-Pentamethyl-1,3-cyclopentadiene
CAS Number:4045-44-7
:
Formula:C₁₀H₁₆
MDL Number:MFCD00001354
Boiling Pt:58 °C/15mmHg
Density:0.87 g/cm³ (25 °C)
Molecular Weight:136.24 g/mol
Flash Pt:44 °C
Storage Temperature:Ambient
EINECS:223-743-4
UN:3295
ADR:3 III
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together