0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
Ethyl(pyridin-2-ylmethyl)amine
Ethyl(pyridin-2-ylmethyl)amine 97%
Ethyl(pyridin-2-ylmethyl)amine 97%
APOSOR315116-5G
:
Apollo Scientific
:
Ethyl(pyridin-2-ylmethyl)amine 97%
APOSOR315116-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
Environmentally Preferable:
:
N-(Pyridin-2-ylmethyl)ethanamine
:
:
136.2 g/mol
:
Ambient
:
51639-58-8
:
C₈H₁₂N₂
:
MFCD07411568