5-Hydroxypiperidin-2-one 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:19365-07-2
:
Formula:C5H9NO2
MDL Number:MFCD02258339
Molecular Weight:115.1305 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together