Order Entry
Ireland
ContactUsLinkComponent
Diethyl-4-oxocyclohexane-1,1-dicarboxylate 97%
Diethyl-4-oxocyclohexane-1,1-dicarboxylate 97%
Catalog # APOSOR22900-1G
Supplier:  Apollo Scientific
CAS Number:  
Diethyl-4-oxocyclohexane-1,1-dicarboxylate 97%
Catalog # APOSOR22900-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Diethyl 4-oxocyclohexane-1,1-dicarboxylate
 :
 :242.27 g/mol
Storage Temperature:Ambient
CAS Number:55704-60-4
Formula:C₁₂H₁₈O₅
MDL Number:MFCD00177942