3-Chloro-4-methoxyphenylboronic acid
Catalog # APOSOR13155-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
About this item
Synonyms: 3-Chloro-4-methoxybenzeneboronic acid, (3-Chloro-4-methoxyphenyl)boronic acid
:
:186.40 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:175883-60-0
Formula:C₇H₈BClO₃
MDL Number:MFCD04039888
Melting Pt:238…242 °C