5-Methyl-2-(2-nitrophenyl)oxazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:52829-70-6
:
Formula:C₁₀H₈N₂O₃
MDL Number:MFCD09801026
Molecular Weight:204.1800 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together