2',4'-Dihydroxy-3'-methylpropiophenone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:63876-46-0
:
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00010818
Melting Pt:124…130 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together