4-Chloro-1H-pyrazole-3-carboxaldehyde
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 4-Chloro-1H-pyrazole-5-carbaldehyde, 4-Chloro-3-formylpyrazole
CAS Number:623570-54-7
:
Formula:C₄H₃ClN₂O
MDL Number:MFCD00110641
Molecular Weight:130.53 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together