Ruthenium 99,95%, sponge
Catalog # APOSIN3058-10G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:10 g
- Seq:3236
- Environmentally Preferable:
Specifications
About this item
:
:101.07 g/mol
Boiling Pt:4900 °C
UN:3089
:GHS02
Melting Pt:2500 °C
Storage Temperature:Ambient
CAS Number:7440-18-8
Formula:Ru
MDL Number:MFCD00011207
Density:12.45 g/cm³ (25 °C)
ADR:4.1 III